Skip to Content
Merck
All Photos(2)

Key Documents

660434

Sigma-Aldrich

Gold nanoparticles

3-5 nm particle size (TEM), dodecanethiol functionalized, 2 % (w/v) in toluene

Synonym(s):

Dodecylthiol functionalized gold nanoparticles

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$1,110.00

$1,110.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$1,110.00

About This Item

MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$1,110.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

description

nanoparticles

form

nanoparticles
solution

concentration

2 % (w/v) in toluene

particle size

3-5 nm (TEM)

density

0.9308 g/mL at 25 °C

functional group

thiol

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCS[Au]

InChI

1S/C12H26S.Au/c1-2-3-4-5-6-7-8-9-10-11-12-13;/h13H,2-12H2,1H3;/q;+1/p-1

InChI key

QMOCMJWMJUUCRE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Dodecanethiol functionalized gold nanoparticles can beused to fabricate microcapsules that can act as contrast agents for computedtomography or gamma imaging and as drug carriers.
Useful for the self-assembly of gold monolayers for nanoelectronics.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shakeel Ahmad Khan et al.
Biomolecules, 10(6) (2020-06-04)
Due to their versatile applications, gold (Au) and silver (Ag) nanoparticles (NPs) have been synthesized by many approaches, including green processes using plant extracts for reducing metal ions. In this work, we propose to use plant extract with active biomedical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service