CLS6511
Corning® Thermowell PCR 96 well plates
96 well plate, Thermowell PCR M plate, polycarbonate, conical bottom, clear, non-sterile, 25/cs
Synonym(s):
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate, PCR plate
Select a Size
$133.00
Select a Size
About This Item
clear polycarbonate plate
conical bottom clear wells
plate format: 96 well Model M
$133.00
Recommended Products
material
clear plate
clear polycarbonate plate
conical bottom clear wells
sterility
non-sterile
feature
lid: no
plate format: 96 well Model M
packaging
pack of 1
case of 25
manufacturer/tradename
Corning 6511
technique(s)
PCR: suitable
size
96 wells
well volume
200 μL
color
clear
suitability
suitable for (PCR applications)
application(s)
agriculture
agriculture
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Thermowell 96 well polycarbonate PCR microplates fit the most commonly used thermal cyclers
- For oil-free operation, plates must be used with sealing tape or a sealing mat and used in conjunction with thermal cyclers that have heated lids
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service