Skip to Content
Merck
All Photos(1)

Key Documents

CLS432090

Extension Collar, for Corning® CoolBox 2XT System

pink

Synonym(s):

Corning Coolcell accessory

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$185.00

$185.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$185.00

About This Item

UNSPSC Code:
41123304
NACRES:
NB.12

$185.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

packaging

pkg of 1

manufacturer/tradename

Corning 432090

L × W × H

21.0 cm × 14.3 cm × 7.0 cm, Internal Dimensions
26.5 cm × 20.0 cm × 7.0 cm, External Dimensions

color

pink

Looking for similar products? Visit Product Comparison Guide

General description

For use with CoolBox XT and CoolBox 2XT to extend the height when using tall tube modules. The collar is magnetized and easily adheres to the unit base.

Legal Information

CoolBox is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service