CLS432025
Corning® CoolBox™, 2XT System
workstation, double capacity, purple
Synonym(s):
Staining, Staining Accessories
Select a Size
$1,450.00
Select a Size
About This Item
$1,450.00
Recommended Products
feature
workstation, double capacity
packaging
pkg of 1
manufacturer/tradename
Corning 432025
size
21.0 cm × 14.3 cm × 13.4 cm, Internal Dimensions
26.5 cm × 20.0 cm × 15.0 cm, External Dimensions
color
purple
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Includes two (2) XT Cooling Cores for 0.5° to 4°C cooling
- Accommodates two (2) sample modules
- Includes a magnetized collar and a 1° to 8°C temperature indicator for visual assurance of proper temperature
- To use with any 30 place Corning CoolRack Tubes Module
- The internal cooling or freezing cores provide 0.5° to 4°C for over 10 hours with lid open and over 16 hours with lid closed
- CoolBox systems may also be used with dry ice to create a compact, portable, snap freezing workstation for bacteria, virus or proteins.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service