CLS3842
Corning® 96 Well Poly-D-Lysine Treated Microplate
flat bottom clear, black polystyrene, bag of 20, sterile, lid
Synonym(s):
96-Well Poly-D-Lysine Plate, Poly-D-Lysine Microplate, Treated Microplate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
black polystyrene
clear wells
flat bottom clear
round wells
sterility
sterile
feature
lid
skirt
packaging
bag of 20
manufacturer/tradename
Corning 3842
size
96 wells
well volume
360 μL
well working volume
75-200 μL
binding type
poly-D-lysine coated surface
Looking for similar products? Visit Product Comparison Guide
Application
Corning® 96 Well Poly-D-Lysine Treated Microplate has been used for purification of recombinant proteins.
Features and Benefits
- Black walls and ultra-thin, clear flat bottoms that enable direct microscopic viewing, minimize background interference in fluorescent assays, and allow readings down to 340nm
- Opaque walls reduce well-to-well crosstalk.
- compati
Legal Information
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Methods in molecular biology (Clifton, N.J.), 1957, 121-137 (2019-03-29)
The many functions of β-arrestin proteins in the desensitization of G-protein-coupled receptors have been well characterized; however, the discovery that this scaffold protein could actually recruit phosphodiesterases (PDEs) to the site of cAMP synthesis changed the way researchers thought about
BMC infectious diseases, 11, 121-121 (2011-05-17)
Countries aiming for malaria elimination require a detailed understanding of the current intensity of malaria transmission within their national borders. National household sample surveys are now being used to define infection prevalence but these are less efficient in areas of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service