CLS3506
Corning® Costar® TC-Treated Multiple Well Plates
size 6 wells, clear, clear polystyrene plate, flat bottom wells, case of 100 (20 Bulk Packs of 5), sterile, lid
Synonym(s):
cell culture plate, multi well plates, multiple well plates, multiwell plates, tissue culture plates
About This Item
Recommended Products
material
clear
clear bottom
clear polystyrene plate
flat bottom wells
round clear wells
sterility
sterile
feature
lid
skirt
packaging
case of 100 (20 Bulk Packs of 5)
manufacturer/tradename
Corning 3506
size
6 wells
well diam.
34.8 mm
well volume
16.8 mL
well working volume
1.9-2.9 mL
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
• Nonreversible lids with condensation rings to reduce contamination
• Individual alphanumerical codes for well identification
• Uniform footprint for ease in stacking
• Treated for optimal cell attachment
• Sterilized by gamma irradiation
• Certified nonpyrogenic
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service