Skip to Content
Merck
All Photos(1)

Documents

CLS3508

Corning® 96 well plates

polystyrene plate, clear wells, PVDF membrane non-sterile, case of 50

Synonym(s):

96 well filter plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
NACRES:
NB.24

product name

Corning® 96 well filter plate, 0.2um PVDF filter, 96 well plate w/ 0.2 PVDF filter membrane, polystyrene, non-sterile, clear, 50/cs

material

PVDF membrane
clear wells
polystyrene plate

sterility

non-sterile

feature

lid: no
membrane: glass fiber filter plate

packaging

case of 50

manufacturer/tradename

Corning 3508

size

96 wells , 96 well filter plate

color

clear

suitability

suitable for

General description

Corning 96 Well Plate, 0.2um PVDF Filter Plate

  • Low-binding hydrophilic PVDF membrane filter plate, 96 well format
  • Over-molding manufacturing process encapsulates individual filter disks in the plate, ensuring 100% integrity of each well
  • Filter plate nozzle design prevents cross-contamination
  • Filter plate is suitable for automated (HT) application due to rigid side wall construction
  • Wide skirt plate design readily accepts bar codes
  • Suitable for: protein kinase, resin-based separation assays; plasmid isolation, DNA purification, or receptor/ligand binding assays
protein kinase, resin-based separation assays; plasmid isolation, DNA purification, or receptor/ligand binding assays

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service