MAGVS2210
Multiscreen® 96 well Plate, hydrophilic PVDF membrane
pore size 0.22 μm, sterile
Synonym(s):
96-Well PVDF Plate, Hydrophilic 96-Well Plate, PVDF 96-Well Plate
About This Item
Recommended Products
material
PVDF membrane (hydrophilic)
acrylic
flat bottom wells
Quality Level
description
Sterile, clear 96-well filter plate with 0.22 um pore size Hydrophilic PVDF membrane for Sample Prep & YAC Culture. Comes in a pack of 10.
sterility
sterile
product line
MultiScreen®
feature
hydrophilic
lid
manufacturer/tradename
MultiScreen®
parameter
50-250 μL sample volume (per well)
technique(s)
activity assay: suitable (protein kinase/phosphatase)
bead-based assay: suitable
ligand binding assay: suitable
receptor binding assay: suitable
filtration area
0.28 cm2
plate size
96 wells
well maximum volume
300 μL
working volume
50-250 μL
matrix
Durapore®
pore size
0.22 μm pore size
binding type
low binding surface
shipped in
ambient
General description
HTS Filter Plates (Plate catalogue numbers that start with MS)
- MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96- and 384-well platforms with a broad selection of membranes and plastics.
- Millipore’s new MultiScreen HTS+ Hi flow filter plate design lessens non-specific binding and reduces variability in both background and signal intensities specifically for biochemical screening assays.
Classic Filter Plates (Plate catalogue numbers that start with MA)
- The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.
Accessory – MultiScreenHTS Vacuum Manifold
- The MultiScreenHTS Vacuum manifold was designed specifically to work with MultiScreenHTS filter plates. The combination results in optimal filtration characteristics and seamless automation integration.
Millicell Assay Plates
- In addition to the traditional assay plate format, filter plates are also available with cell culture trays for growing and assaying suspension and adherent cell lines. Cells grown on filter plates differentiate better than those grown on traditional plastic substrates, resulting in more in vivo-like results This filter plate/tray combination can be used for a variety of cellular assays, including, transport, toxicity and screening assays.
Application
General Filtration
Cell Culture
Physical form
Other Notes
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.
Protocols
Guidelines for various bioassays on the MultiScreen system, which makes it possible to carry out an assay from sample incubation to scintillation counting in a 96-well plate.
Related Content
This provides information about the chemical compatibility of MultiScreen Filter Plates with different reagents.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service