CLS3286
Corning® E-Cube™ culture system kit (without CellCube® Module)
culture system kit, complete, 1/cs
Synonym(s):
lab equipment accessory
Select a Size
$7,130.00
Estimated to ship onMay 29, 2025
Select a Size
About This Item
$7,130.00
Estimated to ship onMay 29, 2025
Recommended Products
feature
components included: includes an oxygenator, medium reservoir, multiple access ports, and all required tubing and fittings
packaging
pack of 1
case of 1 ×
manufacturer/tradename
Corning 3286
General description
- Cells grow in Corning′s parallel-plate CELLCUBE® Module on the same treated polystyrene used in Corning culture vessels.
- The E-Cube System kit consists of an oxygenator, medium reservoir, multiple access ports, and all required tubing and fittings; CELLCUBE Modules must be purchased separately
- The E-Cube System provides a simple method to determine if your cells will grow in the CELLCUBE Module prior to investing in the resources and funding that would be necessary for the larger, automated CELLCUBE System.
Note: Peristaltic pump, CO2 incubator and medium are not provided.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service