Key Documents
CLS3081250LO
Pyrex® large knob top desiccators
replacement desiccator bowl 10.5L large knob top desiccator, 1/cs
Synonym(s):
desiccator bowls, desiccator lower bowls
About This Item
UNSPSC Code:
41104510
Pricing and availability is not currently available.
material
borosilicate glass
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 3081-250LO
suitability
suitable for (general analytical work)
General description
The Pyrex large knob top desiccators are designed for general analytical work. The large sturdy knob makes the cover easy to handle. Warning: Not for use with vacuum applications. Do not subject to heat or pressure.
This is the replacement desiccator bowl which fits Pyrex desiccator CLS3181250.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)