CLS3013
Corning® single use spatulas
microspatula, rounded end/scoop
Synonym(s):
Spatula for Lab Use
All Photos(1)
About This Item
Recommended Products
sterility
sterile
manufacturer/tradename
Corning 3013
L
150 mm
General description
Scoop dimensions:
Maximum scoop width - 0.13 inches (3.302 mm)
scoop can hold < 0.5 mL
Maximum scoop width - 0.13 inches (3.302 mm)
scoop can hold < 0.5 mL
Corning single use spatulas have been designed to save time and provide contamination-free samples. Eliminates need to recycle and sterilize reusable spatulas.
Each spatula is individually packaged, RNase/DNase free, non-pyrogenic and antistatic.
Each spatula is individually packaged, RNase/DNase free, non-pyrogenic and antistatic.
Legal Information
Corning is a registered trademark of Corning, Inc.