This product is packaged in 10/sterile pouches per pack.
Z742387
Levgo® smartSpatula® disposable PP laboratory spatula, Look for the V at the neck™
L 210 mm, white, standard, sterile (EtO), endotoxin free (non-pyrogenic)
Synonym(s):
disposable spatula, endotoxin free spatula, microspatula
Select a Size
$583.00
Select a Size
About This Item
$583.00
Recommended Products
material
polypropylene
white
sterility
sterile (EtO)
feature
endotoxin free (non-pyrogenic)
packaging
pack of 100 ea
case of 500 ea
manufacturer/tradename
LevGo Inc 17271
L
210 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
LevGo spatulas are widely used in commercial, university, diagnostic and pharmaceutical labs worldwide as well as by compounding pharmacists to produce customized medications.
Features and Benefits
- The V design at the neck of the disposable lab spatula is a trademark of LevGo, Inc.
- No Searching, washing or contamination.
- 100% Polypropylene is autoclavable and resistant to dilute acids and bases.
- Polypropylene will not shatter after exposure to liquid nitrogen frozen samples.
- Hollow core uses less plastic than other spatulas and minimizes heat or cold transfer.
- Sustainability – LevGo plants a tree for every case sold.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Can you tell me if the spatula of material "Z742387 L 210 mm, white, sterile, endotoxin free (non-pyrogenic)" are packed per single spatulum or are they packed with more spatula in a package?
1 answer-
Helpful?
-
-
Bonjour, Est il possible d'avoir des échantillons afin de s'assurer que le produit correspond au besoin s'il vous plait ?
1 answer-
Samples of products supplied by other manufacturers may not be available. We kindly ask you to navigate to the link https://www.sigmaaldrich.com/techservice, click on "Product Technical Inquires" under the Products Section with all the required information so that a member of our team can reach out to you to assist further. Thank you.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service