CLS2797
Corning® 96 Well Clear PVC Assay Microplate
round bottom clear, PVC, bag of 25 ×, non-sterile, lid: no
Synonym(s):
cell culture plate, multi well plates, multiple well plates
Select a Size
$245.00
Select a Size
About This Item
round bottom clear
round wells
$245.00
Recommended Products
material
PVC
round bottom clear
round wells
sterility
non-sterile
feature
lid: no
manufacturer/tradename
Corning 2797
packaging
bag of 25 ×
size
96 wells
well volume
250 μL
well working volume
50-150 μL
color
clear
suitability
suitable for (cell cuture)
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Corning® 96 Well Clear PVC Assay Microplate is economical and untreated.
- PVC microplates support solution-based assays, serial dilutions, and general storage applications.
- Well volume of 250 μL; recommended working volume of 50 to 150 μL.
- Lids are not available.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service