CLS9018
Corning® 96 Well EIA/RIA Assay Microplate
flat bottom clear, polystyrene, high binding surface, bag of 25, non-sterile, lid: no
Synonym(s):
96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates
Select a Size
$168.00
Select a Size
About This Item
flat bottom clear
polystyrene
polystyrene
96 wells , 96 well plate
$168.00
Recommended Products
material
clear bottom
flat bottom clear
polystyrene
polystyrene
sterility
non-sterile
feature
lid: no
skirt
packaging
case of 100
bag of 25
manufacturer/tradename
Corning 9018
size
96 wells
96 wells , 96 well plate
well volume
330-360 μL , Note: CLS3366 and CLS3797 have well volume capacities of 330 uL
well working volume
75-200 μL
suitability
suitable for (EIA/RIA; immunoassays)
binding type
high binding surface
Related Categories
General description
- High binding surface is capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions
- Binding capacity is approximately 400 to 500 ng IgG/cm2
- Ideal for immunoassays
- Without lids, lids supplied separately
- Total volume is 360 μL; recommended working volume of 75 to 200 μL
- Conforms to standard microplate footprint and dimensions
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)