Skip to Content
Merck
All Photos(1)

Key Documents

CLS2510

Corning® 1 x 8 Stripwell 96 well plates

Sulfhydryl-BIND surface

Synonym(s):

1 x 8 strip plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
flat bottom clear polystyrene (96-well)
size:
96 wells

material

flat bottom clear polystyrene (96-well)

packaging

plate of 12 strips

manufacturer/tradename

Corning 2510

size

96 wells

well working volume

75-200 μL , for low volume plates volume is 50-125 μL

color

clear

suitability

suitable for (cell cuture)

Looking for similar products? Visit Product Comparison Guide

General description

The Costar 1 × 8 strips are pre-assembled in an "egg crate" style 96 well strip holder. Strips fit in holder only one way, eliminating mis-orientation. If covers are needed, the notched cover fits the fully assembled strip holder.

Legal Information

Corning is a registered trademark of Corning, Inc.
Stripwell is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service