Skip to Content
Merck
All Photos(1)

Key Documents

CLS1500P500

Corning® narrow mouth reagent bottle, reusable

capacity 500 mL, polypropylene, with GL-45 PP screw cap

Synonym(s):

laboratory bottles, plastic bottles, reagent bottles

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$84.90

$84.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
10 EA
$84.90

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

$84.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

cylindrical bottle
narrow-mouth bottle
polypropylene
screw top cap
translucent bottle

feature

cap

packaging

pack of 10 ea

manufacturer/tradename

Corning 1500P-500

H × O.D.

204 mm × 108 mm

capacity

500 mL

fitting

45

Looking for similar products? Visit Product Comparison Guide

General description

These bottles are manufactured from translucent polypropylene. They are supplied with PP screw top caps.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service