Skip to Content
Merck
All Photos(1)

Documents

EJG

93051803, bovine adrenal gland, Endothelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

EJG, 93051803

biological source

bovine adrenal gland

growth mode

Adherent

karyotype

Not specified

morphology

Endothelial

products

Factor VIII

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Bovine adrenal medulla endothelial

Cell Line Description

The bovine cell line was established from collagenase-dissociated, adrenal medulla tissue. The cells display a capillary endothelium morphology with relatively large cell bodies and produce factor VIII:C. Tests for BVDV have shown that the cell line is producing both detectable BVDV antigens and infectious BVDV.

Application

Blood clotting studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. Passage cells every 5 to 7 days.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service