Skip to Content
Merck
All Photos(1)

Key Documents

CYNOM-K1

90071809, cynomolgus monkey skin, Fibroblast

Synonym(s):

CYNOM-K1 Cell Line, Cynom Cell Line K1, Human CYNOM-K1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

CYNOM-K1, 90071809, from cynomolgus monkey skin, Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)

biological source

cynomolgus monkey skin

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

receptors

Not specified

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

Monkey Cynomologus skin

Cell Line Description

Derived from the skin of a Cynomologus monkey embryo.

Application

CYNOM-K1 has been used for the transduction efficiency of hematopoietic cells with chimeric lentiviral vectors.[1]

Culture Medium

EMEM (EBSS) (M2279) + 2 mM L-Glutamine (G7513) + 1% NEAA (M7145) + 10% FBS / FCS (F2442).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6, i.e., seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service