Skip to Content
Merck
All Photos(1)

Documents

RCC4 plus vector alone

3112702, human kidney, Epithelial

Synonym(s):

Human RCC4, RCC4 Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

RCC4 plus vector alone, 03112702

biological source

human kidney

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Renal cell carcinoma cell line RCC4 stably transfected with an empty expression vector, pcDNA3, conferring neomycin resistance.

Cell Line Description

Renal cell carcinoma cell line RCC4 stably transfected with an empty expression vector, pcDNA3, conferring neomycin resistance. This cell line is VHL-deficient i.e. the von Hippel-Lindau tumour suppressor gene product is not expressed. These cells are tumourigenic.

Application

RCC4 plus vector alone is mainly used to study the tumour suppressor gene product function - von Hippel-Lindau tumour suppressor gene. It is also used in cell responses to hypoxia.
  • RCC4 plus vector alone has been used in study involving the effect of in vitro ischemia on canonical hypoxic HIF-1α (Hypoxia Inducible Factor α) stabilization.
  • It has been used in the study involving CMA (chaperone-mediated autophagy)-mediated degradation of HIF1α in the lysosome.
  • It has also been used to study lipodepsipeptide MDN-0066-mediated inhibition of VHL/HIF (Von Hippel-Lindau/hypoxia inducible factor) pathway.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10,12
D13S317: 12
D16S539: 11
D5S818: 12
D7S820: 8.2
THO1: 9.3
TPOX: 11
vWA: 16

Culture Medium

DMEM + 10% FBS + 2 mM Glutamine + G418 0.5mg/ml

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:8 i.e. seeding at 1-3 x 10,000 cells/cm2 using 0.25% Trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service