Key Documents
Z153095
Sigma-Aldrich
Aldrich® jacketed round-bottom flasks
capacity 500 mL
Select a Size
1 EA
$760.00
$760.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$760.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$760.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass
round bottom flask
three-neck
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
500 mL
General description
24/40 joint, with two hose connections. Multifunctional-can be water cooled or heated with hot water or steam.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)