Skip to Content
Merck
All Photos(1)

Key Documents

CAS9D10ARFPP

Sigma-Aldrich

CMV-CAS9D10A-2A-RFP Plasmid

Synonym(s):

CAS9D10A Plasmid

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$299.00

$299.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 4 to 6 weeks.

Request a Bulk Order

Select a Size

Change View
1 EA
$299.00

About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

$299.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 4 to 6 weeks.

Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

packaging

vial of 50 μL

concentration

20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)

Promoter

Promoter name: CMV

reporter gene

RFP

selection

kanamycin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The Cas9-D10A nickase plasmid co-expressing RFP uses the CMV promoter for strong transient expression of Cas9-D10A. Alternate promoters can be substituted by replacement of CMV using MluI and NheI. Also, the Cas9D10A expression plasmids can be linearized using XbaI for T7-based mRNA production.

Application

Functional Genomics
Use of Paired Cas9 Nickase + RFP for:
  • Creation of gene knockouts in multiple cell lines
  • Complete knockout of genes not amenable to RNAi
  • Creation of knock-in cell lines with promoters, fusion tags or reporters integrated into endogenous genes

Components

1 vial containing 1ug of Cas9-D10A Nickase-RFP plasmid.

Please note, this product does not contain any guide RNA sequence. A pair of gRNA plasmids must be purchased separately through the Custom CRISPR paired nickase product tab.

Physical form

2 ug of Sigma Cas9-D10A nickase-RFP plasmid

Other Notes

The type II CRISPR/Cas system from the bacterium Streptococcus pyogenes has been engineered to function in eukaryotic systems using two molecular components: a single Cas9 protein and a non-coding guide RNA (gRNA). The Cas9 endonuclease can be programmed with a single gRNA, directing a DNA double-strand break (DSB) at a desired genomic location. Similar to DSBs induced by zinc finger nucleases (ZFNs), the cell then activates endogenous DNA repair processes, either non-homologous end joining (NHEJ) or homology-directed repair (HDR), to heal the targeted DSB. RFP is co-expressed from the same mRNA as the Cas9 nickase protein via a 2A peptide linkage, enabling tracking of transfection efficiency and enrichment of genome editing activity in cell populations via fluorescence activated cell sorting (FACS).

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

Learn about CRISPR Cas9, what it is and how it works. CRISPR is a new, affordable genome editing tool enabling access to genome editing for all.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service