Skip to Content
Merck
All Photos(1)

Key Documents

C8984

Sigma-Aldrich

Z-Leu-Leu-Leu-fluoromethyl ketone

≥90% (TLC)

Synonym(s):

Z-LLL-FMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H42N3O5F
Molecular Weight:
507.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥90% (TLC)

form

powder

solubility

acetone: 10 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(=O)OCc1ccccc1)C(=O)CF

InChI

1S/C27H42FN3O5/c1-17(2)12-21(24(32)15-28)29-25(33)22(13-18(3)4)30-26(34)23(14-19(5)6)31-27(35)36-16-20-10-8-7-9-11-20/h7-11,17-19,21-23H,12-16H2,1-6H3,(H,29,33)(H,30,34)(H,31,35)/t21-,22-,23-/m0/s1

InChI key

HLXJVMRDGBCZCM-VABKMULXSA-N

Amino Acid Sequence

Z-Leu-Leu-Leu-FMK

Biochem/physiol Actions

Z-Leu-Leu-Leu-fluoromethyl ketone is a proteosome inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tianling Ou et al.
PLoS pathogens, 17(1), e1009212-e1009212 (2021-01-20)
Hydroxychloroquine, used to treat malaria and some autoimmune disorders, potently inhibits viral infection of SARS coronavirus (SARS-CoV-1) and SARS-CoV-2 in cell-culture studies. However, human clinical trials of hydroxychloroquine failed to establish its usefulness as treatment for COVID-19. This compound is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service