Skip to Content
Merck
All Photos(1)

Key Documents

C7499

Sigma-Aldrich

CK-548

≥98% (HPLC)

Synonym(s):

2-(3-Bromophenyl)-3-(5-chloro-2-hydroxyphenyl)-4-thiazolidinone, CK-0993548

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11BrClNO2S
CAS Number:
Molecular Weight:
384.68
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

solubility

DMSO: 24 mg/mL

storage temp.

2-8°C

SMILES string

Oc1ccc(Cl)cc1N2C(SCC2=O)c3cccc(Br)c3

InChI

1S/C15H11BrClNO2S/c16-10-3-1-2-9(6-10)15-18(14(20)8-21-15)12-7-11(17)4-5-13(12)19/h1-7,15,19H,8H2

InChI key

KEGQNJITMFBVAC-UHFFFAOYSA-N

Biochem/physiol Actions

CK-548 inhibits the activity of actin-related protein (Arp)2/3 complex by inserting into the hydrophobic core of Arp3 and altering its conformation.
CK-548 inhibits the activity of actin-related protein (Arp)2/3 complex by inserting into the hydrophobic core of Arp3 and altering its conformation: inhibits

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renu Khasa et al.
Microbial pathogenesis, 149, 104565-104565 (2020-10-16)
Multiple membrane trafficking networks operate in the eukaryotic cell and are hijacked by viruses to establish infection. Recent studied have highlighted that viruses can exploit distinct pathways depending on the cell type. Japanese encephalitis virus (JEV), a neurotropic flavivirus, can
Renu Khasa et al.
The Journal of general virology, 100(2), 176-186 (2018-11-30)
Japanese encephalitis virus (JEV), a mosquito-borne flavivirus, is one of the leading global causes of virus-induced encephalitis. The infectious life-cycle of viruses is heavily dependent on the host membrane trafficking network. Here, we have performed a RNA-interference-based screen using a
Arnat Balabiyev et al.
Molecular biology of the cell, 31(18), 2002-2020 (2020-06-25)
Macrophage fusion resulting in the formation of multinucleated giant cells (MGCs) is a multistage process that requires many adhesion-dependent steps and involves the rearrangement of the actin cytoskeleton. The diversity of actin-based structures and their role in macrophage fusion is
Monika Hartmann et al.
Molecular cancer research : MCR, 13(5), 879-890 (2015-02-06)
Ectodomain cleavage (shedding) of transmembrane proteins by metalloproteases (MMP) generates numerous essential signaling molecules, but its regulation is not totally understood. CD44, a cleaved transmembrane glycoprotein, exerts both antiproliferative or tumor-promoting functions, but whether proteolysis is required for this is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service