Skip to Content
Merck
All Photos(1)

Key Documents

C7275

Sigma-Aldrich

Concanavalin A from Canavalia ensiformis (Jack bean)

Type V, lyophilized powder

Synonym(s):

Con A

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202

type

Type V

form

lyophilized powder

potency

≤64 μg per mL agglutination activity

impurities

Carbohydrate, essentially free
salt, essentially free
≤0.16 wt. % Ca
≤0.23 wt. % Mn

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Con A is not blood group specific but has an affinity for terminal α-D-mannosyl and α-D-glucosyl residues. Ca2+ and Mn2+ ions are required for activity. Con A dissociates into dimers at pH 5.6 or below. Between pH 5.8 and pH 7.0, Con A exists as a tetramer; above pH 7.0 higher aggregates are formed. Con A exhibits mitogenic activity which is dependent on its degree of aggregation. Succinylation results in an active dimeric form which remains a dimer above pH 5.6.

Quality

Highly purified.

Preparation Note

Produced from whole Jack beans (J0250) using the method of Sophianopoulos.

Analysis Note

Where reported, agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, containing Ca2+ and Mn2+ of a 1 mg per mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of human erythrocytes after 1 hr incubation at 25 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guanbin Gao et al.
Traffic (Copenhagen, Denmark), 21(3), 274-296 (2019-12-29)
Protein retention and the transport of proteins and lipids into and out of the Golgi is intimately linked to the biogenesis and homeostasis of this sorting hub of eukaryotic cells. Of particular importance are membrane proteins that mediate membrane fusion
A J Sophianopoulos et al.
Preparative biochemistry, 11(4), 413-435 (1981-01-01)
Concanavalin A (Con A), obtained either commercially or by affinity chromatography, was further purified by incubating at 6-8 hr at pH 3.0-3.2 in 1 M NaCl, 0.08 M glycine and 3 mM each Ca2+ and Mn+2, heat treating at 45
Gustavo Vasen et al.
Communicative & integrative biology, 13(1), 128-139 (2020-10-06)
Chemotactic/chemotropic cells follow accurately the direction of gradients of regulatory molecules. Many G-protein-coupled receptors (GPCR) function as chemoattractant receptors to guide polarized responses. In "a" mating type yeast, the GPCR Ste2 senses the α-cell's pheromone. Previously, phosphorylation and trafficking of
Yeo Reum Park et al.
Journal of microbiology and biotechnology, 28(12), 2106-2112 (2018-11-02)
Concanavalin A (ConA) interacts with carbohydrates as a lectin, and recent reports proposed its application for detecting a diversity of viruses and pathogens. Structural studies have detailed the interaction between ConA and carbohydrates and the metal coordination environment with manganese
Luis J García-Rodríguez et al.
Current biology : CB, 29(9), 1536-1544 (2019-04-23)
For proper chromosome segregation in mitosis, sister kinetochores must interact with microtubules from opposite spindle poles (chromosome bi-orientation) [1, 2]. To promote bi-orientation, Aurora B kinase disrupts aberrant kinetochore-microtubule interactions [3-6]. It has long been debated how Aurora B halts this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service