Skip to Content
Merck
All Photos(2)

Documents

C7170

Sigma-Aldrich

Chitin from shrimp shells

practical grade, powder

Synonym(s):

Poly-(1→4)-β-N-acetyl-D-glucosamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

shrimp (Pandalus borealis)

grade

practical grade

form

powder

color

white to beige

storage temp.

room temp

SMILES string

CC(=O)N[C@@H]1[C@H](O)O[C@@H](CO)[C@H](O)[C@H]1O

InChI

1S/C8H15NO6/c1-3(11)9-5-7(13)6(12)4(2-10)15-8(5)14/h4-8,10,12-14H,2H2,1H3,(H,9,11)/t4?,5?,6?,7?,8-/m1/s1

InChI key

OVRNDRQMDRJTHS-WTZNIHQSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chitin from shrimp shells is suitable for analysis of chitinase. It is a polysaccharide composed of N-acetyl-D-glucomasine that can also be used for the manufacture of glucosamine and chitosan.
Chitin from shrimp shells, a long-chain polymer of a β-1,4 linked N-acetylglucosamine residues, may be used to study its physicochemical properties and role in the formation of arthropod exoskeletons. It may be used to identify and characterize chitinase(s) and chitin deacetylase(s) and as a control to study the structure of chitins from other species. It may be used in chitin asthma and allergy research. Chitin may be used as a viscosity/thickening agent or binder.

Quality

Requires purification prior to use as a substrate for chitinase.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xinxin He et al.
Marine drugs, 18(3) (2020-03-04)
Chitinase, as one of the most important extracellular enzymes in the marine environment, has great ecological and applied values. In this study, two chitinases (Chi1557 and Chi4668) with 97.33% amino acid sequences identity were individually found in Vibrio rotiferianus and
Shikha Singh et al.
Polymers, 12(3) (2020-03-28)
The crystalline phase of poly(lactic acid) (PLA) has crucial effects on its own properties and nanocomposites. In this study, the isothermal crystallization of PLA, triethyl citrate-plasticized PLA (PLA-TEC), and its nanocomposite with chitin nanocrystals (PLA-TEC-ChNC) at different temperatures and times
Kati Seitz et al.
International journal of molecular sciences, 21(15) (2020-08-01)
Mitogen-activated protein kinases (MAPKs) are key regulators of numerous biological processes in plants. To better understand the mechanisms by which these kinases function, high resolution measurement of MAPK activation kinetics in different biological contexts would be beneficial. One method to
Sara A Burt et al.
PloS one, 9(4), e93414-e93414 (2014-04-03)
The formation of biofilm by bacteria confers resistance to biocides and presents problems in medical and veterinary clinical settings. Here we report the effect of carvacrol, one of the major antimicrobial components of oregano oil, on the formation of biofilms
Qiang Yan et al.
Applied microbiology and biotechnology, 101(20), 7567-7578 (2017-09-09)
Microbial utilization of chitin, a potential renewable biomass feedstock, is being pursued as a means of developing novel consolidated bioprocessing for the production of chemicals. Serratia marcescens is a gram-negative bacterium that is known for its chitinolytic capability and as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service