Skip to Content
Merck
All Photos(2)

Key Documents

C6456

Sigma-Aldrich

9-Cyano-N,N,N′-triethylpyronine-N′-caproic acid N4-(maleimidoethyl)piperazide chloride

≥80% purity (HPLC), solid

Synonym(s):

NIR-2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C36H45N6O4Cl
CAS Number:
Molecular Weight:
661.23
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

9-Cyano-N,N,N′-triethylpyronine-N′-caproic acid N4-(maleimidoethyl)piperazide chloride, ≥80% (HPLC)

Quality Level

Assay

≥80% (HPLC)

form

solid

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

[Cl-].CCN(CC)c1ccc2c(C#N)c3ccc(cc3[o+]c2c1)N(CC)CCCCCC(=O)N4CCN(CC4)CCN5C(=O)C=CC5=O

InChI

1S/C36H45N6O4.ClH/c1-4-39(5-2)27-11-13-29-31(26-37)30-14-12-28(25-33(30)46-32(29)24-27)40(6-3)17-9-7-8-10-34(43)41-21-18-38(19-22-41)20-23-42-35(44)15-16-36(42)45;/h11-16,24-25H,4-10,17-23H2,1-3H3;1H/q+1;/p-1

InChI key

SWBDBUAWUNWTAS-UHFFFAOYSA-M

Application

Fluorescent protein label.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service