Skip to Content
Merck
All Photos(1)

Documents

C1875

Sigma-Aldrich

Decanoic acid

≥98.0%

Synonym(s):

1-Nonanecarboxylic acid, Acid C10, C10:0, Capric acid, NSC 5025

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8COOH
CAS Number:
Molecular Weight:
172.26
Beilstein:
1754556
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

plant

vapor pressure

15 mmHg ( 160 °C)

Assay

≥98.0%

form

crystals

bp

268-270 °C (lit.)

mp

27-32 °C (lit.)

density

0.893 g/mL at 25 °C (lit.)

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCC(O)=O

InChI

1S/C10H20O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3,(H,11,12)

InChI key

GHVNFZFCNZKVNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Decanoic acid is a ten-carbon, saturated fatty acid. It is present in palm kernel, coconut fat and in milk fat.

Application

Decanoic acid is used in the preparation of ammonium decanoate (a surfactant) and for the preparation of disperse phase.

Biochem/physiol Actions

Decanoic acid is helpful in the attenuation of oxidative stress. Decanoic acid in ketogenic diet is involved in mitochondrial biogenesis thereby enhancing the citrate synthase and complex I activity of electron transport chain.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

296.6 °F - closed cup

Flash Point(C)

147 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colloidal processing of macroporous TiO2 materials for photocatalytic water treatment
Natoli A, et al.
Journal of the American Ceramic Society. American Ceramic Society, 95(2), 502-508 (2012)
Function of capric acid in cyclophosphamide-induced intestinal inflammation, oxidative stress, and barrier function in pigs
Lee SI and Kang KS
Scientific reports, 7(1), 16530-16530 (2017)
Principles of Orthomolecularism (2004)
Lexicon of lipid nutrition (IUPAC Technical Report)
Beare-Rogers JL, et al.
Pure and Applied Chemistry. Chimie Pure Et Appliquee, 73(4), 685-744 (2001)
A descriptive force-balance model for droplet formation at microfluidic Y-junctions
Steegmans MLJ, et al.
AIChE Journal, 56(10), 2641-2649 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service