Skip to Content
Merck
All Photos(1)

Documents

BAF198560250

Poxygrid® Centrifuge Bottle Rack

Holds 6 x 250 mL bottles, round bottom conical bottom

Synonym(s):

bottle storage

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.22

material

epoxy-coated (steel)

feature

Holds 6 x 250 mL bottles

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F19856-0250

Looking for similar products? Visit Product Comparison Guide

General description

  • Each well is 60mm square to hold bottles.
  • Epoxy coated steel wire is chemical and corrosion resistant and steam autoclavable at 121ºC (250ºF).
  • Organize, transport and store conical and round bottom centrifuge bottles.

Features and Benefits

  • Holds large conical and round-bottom centrifuge tubes that cannot stand up by themselves such as Falcon®, Corning®, Nunc®, and Kimble®).
  • Heat-cured epoxy coating, unaffected by organic solvents, salts, weak acids and weak alkalis.

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.
KIMBLE is a registered trademark of DWK Life Sciences
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Poxygrid is a registered trademark of SP Scienceware, a division of SP

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service