Skip to Content
Merck
All Photos(2)

Documents

B950-05

Sigma-Aldrich

Bovine Renal Artery Smooth Muscle Cells: BRASMC (Cryovial)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

bovine renal artery (normal, tunica intima and media)

Quality Level

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 60

morphology

smooth muscle

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

renal disease; cardiovascular diseases

shipped in

dry ice

storage temp.

−196°C

Related Categories

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Bovine Renal Artery Smooth Muscle Cells (BRASMC) are derived from tunica intima and tunica media of normal bovine renal arteries. They are cryopreserved at second passage and can be cultured and propagated at least 12 population doublings. Studies show cyclosporine-induced renal artery smooth muscle contraction is associated with increases in the phosphorylation of specific contractile regulatory proteins1. The renal artery smooth muscle is uniquely refractory to angiotensin stimulation and may be protective in the role of the kidney in regulating blood pressure through the renin-angiotensin system2.

Beall, A. et al, Biochem Biophys Acta., 1449(1):41-9 (1999)
Whitney, E.G. et al, J. Surg. Res. 61(2):307-10 (1996)

Products are for research use only. They are not intended for human, animal, or diagnostic application.
Animal tissues were USDA inspected.

Cell Line Origin

Artery

Application

blood flow to kidneys

Components

Bovine Smooth Muscle Cell Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Bovine Smooth Muscle Cell Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 12 doublings

Subculture Routine

Please refer to the BRASMC Culture Protocol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service