Skip to Content
Merck
All Photos(1)

Key Documents

SRP3202

Sigma-Aldrich

Granzyme B from mouse

recombinant, expressed in Hi-5 Insect cells, ≥97% (SDS-PAGE), ≥97% (HPLC), suitable for cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$453.00

$453.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.


Select a Size

Change View
10 μG
$453.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$453.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

biological source

mouse

recombinant

expressed in Hi-5 Insect cells

Assay

≥97% (HPLC)
≥97% (SDS-PAGE)

form

lyophilized

mol wt

28.9 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... GZMB(14939)

General description

Granzyme B is a cysteine protease found in the cytoplasmic granules of cytolytic T lymphocytes (CTL) and natural killer (NK) cells. Granzyme B is required for the induction of target cell lysis, which occurs as part of cell mediated immune responses, and can activate apoptosis in target cells by both caspase dependent and caspase independent mechanisms. Proteolytic cleavage of substrates by Granzyme B takes place primarily after aspartic acid residues. Recombinant murine Granzyme B is a glycosylated 227 amino acid protein, comprising the mature active portion of the murine Granzyme B precursor. The apparent molecular weight is 28.9 kDa by mass spectrometry.

Biochem/physiol Actions

Granzyme B is a cysteine protease found in the cytoplasmic granules of cytolytic T lymphocytes (CTL) and natural killer (NK) cells. Granzyme B is required for the induction of target cell lysis, which occurs as part of cell mediated immune responses, and can activate apoptosis in target cells by both caspase dependent and caspase independent mechanisms. Proteolytic cleavage of substrates by Granzyme B takes place primarily after aspartic acid residues. Recombinant murine Granzyme B is a glycosylated 227 amino acid protein, comprising the mature active portion of the murine Granzyme B precursor. The apparent molecular weight is 28.9 kDa by mass spectrometry.

Sequence

IIGGHEVKPH SRPYMALLSI KDQQPEAICG GFLIREDFVL TAAHCEGSII NVTLGAHNIK EQEKTQQVIP MVKCIPHPDY NPKTFSNDIM LLKLKSKAKR TRAVRPLNLP RRNVNVKPGD VCYVAGWGRM APMGKYSNTL QEVELTVQKD RECESYFKNR YNKTNQICAG DPKTKRASFR GDSGGPLVCK KVAAGIVSYG YKDGSPPRAF TKVSSFLSWI KKTMKSS

Physical form

Lyophilized from 1x PBS.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Granzymes at a glance.
Michael Bots et al.
Journal of cell science, 119(Pt 24), 5011-5014 (2006-12-13)
Katherine M Horvath et al.
Respiratory research, 12, 102-102 (2011-08-06)
Modified function of immune cells in nasal secretions may play a role in the enhanced susceptibility to respiratory viruses that is seen in smokers. Innate immune cells in nasal secretions have largely been characterized by cellular differentials using morphologic criteria
Inka Albrecht et al.
The Journal of clinical investigation, 125(12), 4612-4624 (2015-11-10)
Mutations of the gene encoding four-and-a-half LIM domain 1 (FHL1) are the causative factor of several X-linked hereditary myopathies that are collectively termed FHL1-related myopathies. These disorders are characterized by severe muscle dysfunction and damage. Here, we have shown that

Questions

  1. Hello, Is this enzyme in its active form? What are the best condition for resuspension? Thank you.

    1 answer
    1. As provided, this is the active form of mouse Granzyme B and not the proform. Activity level is determined by the ability to cleave a synthetic chromogenic Granzyme B substrate. For resuspension and solubility: Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8 °C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (e.g., 0.1% BSA) and store in working aliquots at -20 °C to -80 °C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service