Skip to Content
Merck
All Photos(1)

Key Documents

B9276

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl octanoate

≥98%

Synonym(s):

5-Bromo-4-chloro-3-indolyl caprylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$186.00

$186.00


Estimated to ship onApril 24, 2025



Select a Size

Change View
5 MG
$186.00

About This Item

Empirical Formula (Hill Notation):
C16H19BrClNO2
CAS Number:
Molecular Weight:
372.68
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

$186.00


Estimated to ship onApril 24, 2025


Quality Level

Assay

≥98%

form

solid

storage temp.

−20°C

SMILES string

CCCCCCCC(=O)Oc1c[nH]c2ccc(Br)c(Cl)c12

InChI

1S/C16H19BrClNO2/c1-2-3-4-5-6-7-14(20)21-13-10-19-12-9-8-11(17)16(18)15(12)13/h8-10,19H,2-7H2,1H3

InChI key

QTWMXGHFXBQNEJ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service