Documents
B8032
Sigma-Aldrich
Nalgene® bottles, style 2006
capacity 30 mL
Synonym(s):
nalgene bottle, nalgene bottles
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
polypropylene cap
polypropylene copolymer bottle (PPCO)
round translucent bottle
Joint
Joints threaded neck (20)
description
Autoclavable
feature
with cap
packaging
pack of 12 ea
manufacturer/tradename
Nalgene® 2006-0001
bottle diam. × H
34 mm × 61 mm
capacity
30 mL
General description
Boston style (round) narrow-mouth bottles. Same profile as style 2002.
Features and Benefits
  • Nalgene® narrow-mouth bottles are ideal for long-term storage.
  • Made from polypropylene copolymer (PPCO) material for excellent sample clarity.
  • Highly chemical resistant to most acids, bases, and alcohols.
  • Autoclavable - with and without contents.
  • Laboratory-grade plastic materials for dependably low leachables and extractables.
  • Liner less caps ensures leak-proof seal for sample protection.
  • Light-weight - enables easy lifting and transporting.
  • Avoid UV exposure.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries