Skip to Content
Merck
All Photos(1)

Key Documents

B6777

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt

BioReagent, ≥99%, powder, suitable for molecular biology

Synonym(s):

5-Bromo-4-chloro-3-indoxyl phosphate p-toluidine salt, BCIP® p-toluidine salt, X-phosphate p-toluidine salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$170.00
500 MG
$599.00

$170.00


Estimated to ship onMay 24, 2025



Select a Size

Change View
100 MG
$170.00
500 MG
$599.00

About This Item

Empirical Formula (Hill Notation):
C8H6BrClNO4P · C7H9N
CAS Number:
Molecular Weight:
433.62
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

$170.00


Estimated to ship onMay 24, 2025


Product Name

5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt, BioReagent, for molecular biology, powder, ≥99%

grade

Molecular Biology
for molecular biology

product line

BioReagent

Assay

≥99%

form

powder

solubility

DMF: 20 mg/mL
H2O: insoluble

foreign activity

Protease, none detected

storage temp.

−20°C

SMILES string

Cc1ccc(N)cc1.OP(O)(=O)Oc2c[nH]c3ccc(Br)c(Cl)c23

InChI

1S/C8H6BrClNO4P.C7H9N/c9-4-1-2-5-7(8(4)10)6(3-11-5)15-16(12,13)14;1-6-2-4-7(8)5-3-6/h1-3,11H,(H2,12,13,14);2-5H,8H2,1H3

InChI key

QEIFSLUFHRCVQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
16670B6149B8503
solubility

DMF: 20 mg/mL, H2O: insoluble

solubility

-

solubility

H2O: 20 mg/mL, DMF: insoluble

solubility

DMF: 20 mg/mL, clear to slightly hazy, yellow

grade

for molecular biology

grade

-

grade

-

grade

-

Quality Level

200

Quality Level

-

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

Application

For the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry.
BCIP is provided in two salt forms: the disodium salt which is soluble in water and the p-toluidine form which is soluble in dimethylformamide. These salt forms may be used to prepare a stock solution that in combination with NBT and a reaction buffer, form a substrate solution for alkaline phosphatase. This substrate system, when incubated with alkaline phosphatase, produces an insoluble NBT diformazan product that is easily observable with its purple color. For added convenience, a mixture of BCIP and NBT is provided in an easily dissolvable tablet form or as a ready-to-use liquid.
For the colorimetric detection of alkaline phosphatase-labeled molecules.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Substrates

Histochemical substrate for alkaline phosphatase.

Legal Information

BCIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karuppiah Muthumani et al.
Science translational medicine, 7(301), 301ra132-301ra132 (2015-08-21)
First identified in 2012, Middle East respiratory syndrome (MERS) is caused by an emerging human coronavirus, which is distinct from the severe acute respiratory syndrome coronavirus (SARS-CoV), and represents a novel member of the lineage C betacoronoviruses. Since its identification
Fernando Cartón-García et al.
Scientific reports, 5, 12312-12312 (2015-07-24)
Inherited MYO5B mutations have recently been associated with microvillus inclusion disease (MVID), an autosomal recessive syndrome characterized by intractable, life-threatening, watery diarrhea appearing shortly after birth. Characterization of the molecular mechanisms underlying this disease and development of novel therapeutic approaches
Dorota Rybaczek et al.
Histochemistry and cell biology, 135(3), 263-280 (2011-02-25)
Here, we demonstrate that in HeLa cells, Ser317 of Chk1 undergoes phosphorylation in response to replication stress induced by hydroxyurea. We also demonstrate the existence of constitutive (interphase and mitotic) Chk1 kinase phosphorylation, the translocation of its phosphorylated form from
Kum C Hiong et al.
The Journal of experimental biology, 218(Pt 23), 3717-3728 (2015-10-10)
This study aimed to sequence and characterize two pro-coagulant genes, coagulation factor II (f2) and fibrinogen gamma chain (fgg), from the liver of the African lungfish Protopterus annectens, and to determine their hepatic mRNA expression levels during three phases of
Daniel Steil et al.
Journal of lipid research, 56(12), 2322-2336 (2015-10-16)
Shiga toxins (Stxs) are produced by enterohemorrhagic Escherichia coli (EHEC), which cause human infections with an often fatal outcome. Vero cell lines, derived from African green monkey kidney, represent the gold standard for determining the cytotoxic effects of Stxs. Despite

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service