Skip to Content
Merck
All Photos(1)

Documents

B5681

Sigma-Aldrich

Bay 11-7085

≥98% (HPLC), solid

Synonym(s):

(E)-3-(4-t-Butylphenylsulfonyl)-2-propenenitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15NO2S
CAS Number:
Molecular Weight:
249.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: soluble >25 mg/mL
H2O: insoluble

SMILES string

CC(C)(C)c1ccc(cc1)S(=O)(=O)\C=C\C#N

InChI

1S/C13H15NO2S/c1-13(2,3)11-5-7-12(8-6-11)17(15,16)10-4-9-14/h4-8,10H,1-3H3/b10-4+

InChI key

VHKZGNPOHPFPER-ONNFQVAWSA-N

Gene Information

human ... NFKB1(4790)

Application

Bay 11-7085 has been used as:
  • nuclear factor-κB inhibitor in Panc-1 cells, macrophages
  • inhibitory κB kinase IKK inhibitor in human astrocytoma cells U87MGs
  • phospho-p65 inhibitor human monocyte cell line THP-1

Biochem/physiol Actions

Bay 11-7085 inactivates peroxisome proliferator-activated receptors γ (PPAR-γ) and increases the microtubule-associated proteins 1A/1B light chain 3B (LC3B), a marker of autophagy resulting in apoptosis in human synovial fibroblasts. It promotes apoptosis in pancreatic carcinoma and may serve as radiotherapy-sensitizing drug. BAY 11-7085 decreases B-cell lymphoma 2 (Bcl-2) by inhibiting pI-κBα (inhibitor of nuclear factor κ B kinase subunit α
Inhibits NF-κB activated expression of ICAM-1, VCAM-1, E-selectin, IL-6 and IL-8.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qin Yan et al.
PLoS pathogens, 10(6), e1004149-e1004149 (2014-06-06)
Using A/J mice, which are susceptible to Staphylococcus aureus, we sought to identify genetic determinants of susceptibility to S. aureus, and evaluate their function with regard to S. aureus infection. One QTL region on chromosome 11 containing 422 genes was
The role of IKKbeta in Venezuelan equine encephalitis virus infection
Amaya M, et al.
Testing, 9(2), e86745-e86745 (2014)
Maturational differences in lung NF-kappaB activation and their role in tolerance to hyperoxia
Yang G, et al.
The Journal of Clinical Investigation, 114(5), 669-678 (2004)
Impact of Bay 11-7085 on radiosensitivity of pancreatic cancer cell line Sw1990
Shu C, et al.
International Journal of Clinical and Experimental Medicine, 9(6), 9322-9326 (2016)
BAY 11-7085 induces glucocorticoid receptor activation and autophagy that collaborate with apoptosis to induce human synovial fibroblast cell death
Relic B, et al.
Oncotarget, 7(17), 23370-23370 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service