Skip to Content
Merck
All Photos(1)

Key Documents

D5878

Sigma-Aldrich

Digitoxin

≥92% (HPLC), powder

Synonym(s):

5β,20(22)-Cardenolide-3β,14-diol-3-(O-2,6-dideoxy-β-D-ribo-hexopyranosyl-[1→4]-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-[1→4]-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy, Digitoxoside, Lanatoxin

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$137.00
500 MG
$229.00
1 G
$393.00
5 G
$1,350.00
10 G
$2,540.00

$137.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$137.00
500 MG
$229.00
1 G
$393.00
5 G
$1,350.00
10 G
$2,540.00

About This Item

Empirical Formula (Hill Notation):
C41H64O13
CAS Number:
Molecular Weight:
764.94
Beilstein:
76678
EC Number:
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

$137.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥92% (HPLC)

form

powder

mp

240 °C (dec.) (lit.)

solubility

chloroform: soluble

SMILES string

[H][C@@]1(C[C@H](O)[C@H](O[C@@]2([H])C[C@H](O)[C@H](O[C@@]3([H])C[C@H](O)[C@H](O)[C@@H](C)O3)[C@@H](C)O2)[C@@H](C)O1)O[C@H]4CC[C@@]5(C)[C@]([H])(CC[C@]6([H])[C@]5([H])CC[C@]7(C)[C@]([H])(CC[C@]67O)C8=CC(=O)OC8)C4

InChI

1S/C41H64O13/c1-20-36(46)29(42)16-34(49-20)53-38-22(3)51-35(18-31(38)44)54-37-21(2)50-33(17-30(37)43)52-25-8-11-39(4)24(15-25)6-7-28-27(39)9-12-40(5)26(10-13-41(28,40)47)23-14-32(45)48-19-23/h14,20-22,24-31,33-38,42-44,46-47H,6-13,15-19H2,1-5H3/t20-,21-,22-,24-,25+,26-,27+,28-,29+,30+,31+,33+,34+,35+,36-,37-,38-,39+,40-,41+/m1/s1

InChI key

WDJUZGPOPHTGOT-XUDUSOBPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Digitoxin is a cardiac glycoside (CG). It is a cardiotonic drug, which exhibits cardiac[1] and anti-cancer properties.[2] Digitoxin inhibits nuclear factor kappa B (NF-κB) signaling.[3] It is used to treat congestive heart failure and cardiac arrhythmia.[1] Digitoxin prevents microtubule formation.[4]

Application

Digitoxin has been used as a labeled drugs for binding sites on human serum albumin (HSA).[5][6] It has also been used to test its anti-transmissible gastroenteritis virus (TGEV) activity.[7]

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Digitoxin for airway inflammation in cystic fibrosis: preliminary assessment of safety, pharmacokinetics, and dose finding
Zeitlin PL, et al.
Annals of the American Thoracic Society, 14(2), 220-229 (2017)
Biointeractions of herbicide atrazine with human serum albumin: UV-Vis, fluorescence and circular dichroism approaches
Zhu M, et al.
International Journal of Environmental Research and Public Health, 15(1), 116-116 (2018)
Identification of anti-viral activity of the cardenolides, Na+/K+-ATPase inhibitors, against porcine transmissible gastroenteritis virus
Yang CW, et al.
Toxicology and Applied Pharmacology, 337, 129-137 (2017)
Digitoxin and its analogs as novel cancer therapeutics
Elbaz HA, et al.
Experimental Hematology & Oncology, 1(1), 4-4 (2012)
Comparative interactions of dihydroquinazolin derivatives with human serum albumin observed via multiple spectroscopy
Wang Y, et al.
Applied Sciences, 7(2), 200-200 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service