Skip to Content
Merck
All Photos(1)

Key Documents

SMB00271

Sigma-Aldrich

Chebulinic acid

≥85% (LC/MS-ELSD)

Synonym(s):

Eutannin

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$592.00

$592.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$592.00

About This Item

Empirical Formula (Hill Notation):
C41H32O27
CAS Number:
Molecular Weight:
956.68
UNSPSC Code:
12352205
NACRES:
NA.25

$592.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥85% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@@H]2[C@H](O[C@H]([C@H](OC(=O)c6c7c(c(c(c6)O)O)OC(=O)[C@H]([C@H]7[C@@H](C1=O)CC(=O)O)O)[C@H]2OC(=O)c5cc(c(c(c5)O)O)O)OC(=O)c4cc(c(c(c4)O)O)O)COC(=O)c3cc(c(c(c3)O)O)O

InChI

1S/C41H32O27/c42-15-1-10(2-16(43)26(15)51)35(56)62-9-22-31-33(66-36(57)11-3-17(44)27(52)18(45)4-11)34(41(63-22)68-37(58)12-5-19(46)28(53)20(47)6-12)67-38(59)13-7-21(48)29(54)32-25(13)24(30(55)40(61)65-32)14(8-23(49)50)39(60)64-31/h1-7,14,22,24,30-31,33-34,41-48,51-55H,8-9H2,(H,49,50)/t14-,22+,24-,30-,31+,33-,34+,41-/m0/s1

InChI key

YGVHOSGNOYKRIH-FJPMMHPYSA-N

General description

Natural product derived from plant source.

Biochem/physiol Actions

Chebulinic acid is polyphenol that has shown antihypertensive, and anti-oxidant properties. Its antihypertensive effects may be due to decreased cardiac output from reduced left ventricular contraction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyun-Sun Lee et al.
Journal of ethnopharmacology, 131(3), 567-574 (2010-07-28)
The aqueous extract of Terminalia chebular fruits was reported to have anti-hyperglycemia and anti-diabetic complication effects. The present study therefore investigated the protective mechanism of chebulic acid, a phenolcarboxylic acid compound isolated from the ripe fruits of Terminalia chebula against
Zong-Chun Yi et al.
Cancer letters, 242(1), 77-87 (2005-12-13)
Tellimagrandin I and chebulinic acid, two hydrolysable tannins, have been shown to exert anti-tumor properties. Dysfunctional gap junctional communication (GJIC) has been recognized as being involved in carcinogenesis. The human cervical carcinoma HeLa cells have been reported to be deficient
Y Y Guan et al.
Clinical and experimental pharmacology & physiology, 23(8), 747-750 (1996-08-01)
1. The effects of chebulinic acid, which has been shown to elicit blood pressure lowering effect in rats, on aortic vascular contraction as well as cardiac contraction were studied in rats. 2. Chebulinic acid had no effect on KCl-induced aortic
Zong-Chun Yi et al.
Toxicology in vitro : an international journal published in association with BIBRA, 23(4), 667-673 (2009-03-31)
The effects of two polyphenols, chebulinic acid and tellimagrandin I, on DNA strand breaks mediated by H(2)O(2)/Cu(II), hydroquinone (HQ)/Cu(II) and H(2)O(2)/Fe(II) in pBR322 plasmid DNA and genomic DNA of cultured MRC-5 human embryo lung fibroblasts were examined. The results demonstrated
Kai Lu et al.
PloS one, 7(8), e43934-e43934 (2012-09-01)
Triphala churna (THL) is a combination of three fruits that has been used for many years in India for the treatment of various diseases. There are now reports which indicate that THL can inhibit growth of malignant tumors in animals.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service