Skip to Content
Merck
All Photos(1)

Documents

B3269

Sigma-Aldrich

Bromphenol Blue–Xylene Cyanole Dye solution

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

product name

Bromphenol Blue–Xylene Cyanole Dye solution, lyophilized powder

form

lyophilized powder

Quality Level

color

dark blue

pH range

8-8.6

solubility

water: 0.2 vial/mL, blue

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

General description

Bromophenol Blue–Xylene Cyanol Dye solution is a tracking dye for nucleic acid electrophoresis in agarose or polyacrylamide gels including DNA sequencing.

Application

Bromophenol Blue–Xylene Cyanol Dye solution has been used in the formamide loading buffer for the size purification of ligated RNA from polyacrylamide gel.

Reconstitution

Add 5 mL water to the vial to dissolve. Reconstituted solution contains 0.5% bromphenol blue and 0.5% xylene cyanole. Dilute 1:10 for nucleic acid electrophoresis in agarose or polyacrylamide gels or add 4 parts formamide to 1 part dye solution for DNA sequencing.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genomic sequencing.
Church GM and Walter G
Proceedings of the National Academy of Sciences of the USA, 81.7, 1991-1995 (1984)
CLIP: construction of cDNA libraries for high-throughput sequencing from RNAs cross-linked to proteins in vivo.
Wang Z, et al.
Methods, 48, 287-293 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service