Skip to Content
Merck
All Photos(1)

Documents

B0543

Sigma-Aldrich

Nunc® Storage Boxes for Nunc CryoTubes®

Synonym(s):

cryogenic storage box, cryotube storage box, cryovial storage box

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123304
NACRES:
NB.22

material

cardboard

feature

Holds 81 x 1-1.8 mL tubes

packaging

pack of 6

manufacturer/tradename

Nunc 378220

technique(s)

cryopreservation: suitable

external L × W × H

132 mm × 132 mm × 51 mm

color

white

suitability

suitable for (cryotube/cryovial storage)

Looking for similar products? Visit Product Comparison Guide

Legal Information

CryoTube is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service