Key Documents
AXYP384240SQCSI
Corning® Axygen® Deep Well Microplate
indiv. wrapped, size 384 wells, working volume 240 μL, pkg of 50 ea, sterile
Synonym(s):
Corning cell culture plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
square bottom clear polypropylene wells
maximum volume:
250 μL
size:
384 wells
working volume:
240 μL
Pricing and availability is not currently available.
material
square bottom clear polypropylene wells
sterility
sterile
packaging
pkg of 50 ea
manufacturer/tradename
Axygen Inc P-384-240SQ-C-SI
Corning
maximum volume
250 μL
size
384 wells
working volume
240 μL
binding type
non-treated surface
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)