Skip to Content
Merck
All Photos(2)

Documents

AV47083

Sigma-Aldrich

Anti-KIAA0319 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-DLX2, Anti-DYLX2, Anti-DYX2, Anti-KIAA0319, Anti-MGC176717

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KIAA0319(9856)

Immunogen

Synthetic peptide directed towards the N terminal region of human KIAA0319

Application

Anti-KIAA0319 antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/mL. It is also useful for immunohistochemistry at a concentration of 4-8μg/mL.

Biochem/physiol Actions

KIAA0319 gene encodes a single-pass type I membrane protein primarily expressed in brain cortex, putamen, amygdala, hippocampus and cerebellum. It plays a crucial role in regulating neuronal migration and cell adhesion and hence facilitates the development of cerebral cortex. KIAA0319 may also regulate the growth and differentiation of dendrites.

Sequence

Synthetic peptide located within the following region: EEMSEYSDDYRELEKDLLQPSGKQEPRGSAEYTDWGLLPGSEGAFNSSVG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Paracchini et al.
Human molecular genetics, 15(10), 1659-1666 (2006-04-08)
Dyslexia is one of the most prevalent childhood cognitive disorders, affecting approximately 5% of school-age children. We have recently identified a risk haplotype associated with dyslexia on chromosome 6p22.2 which spans the TTRAP gene and portions of THEM2 and KIAA0319.
Veronica J Peschansky et al.
Cerebral cortex (New York, N.Y. : 1991), 20(4), 884-897 (2009-08-15)
We investigated the postnatal effects of embryonic knockdown and overexpression of the candidate dyslexia gene homolog Kiaa0319. We used in utero electroporation to transfect cells in E15/16 rat neocortical ventricular zone with either 1) small hairpin RNA (shRNA) vectors targeting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service