Skip to Content
Merck
All Photos(1)

Key Documents

AV09007

Sigma-Aldrich

Anti-RGS3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-C2PA, Anti-FLJ20370, Anti-FLJ31516, Anti-FLJ90496, Anti-PDZ-RGS3, Anti-RGP3, Anti-Regulator of G-protein signaling 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
rat, bovine, pig, human, horse, dog, mouse
citations:
3
technique(s):
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

101 kDa

species reactivity

rat, bovine, pig, human, horse, dog, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RGS3(5998)

Immunogen

Synthetic peptide directed towards the C terminal region of human RGS3

Application

Anti-RGS3 antibody produced in rabbit is suitable for western blotting at a concentration of 0.25 μg/ml.

Biochem/physiol Actions

Regulator of G-protein signaling-3 (RGS3) accelerates the GTPase activity of G(i) and G(q) subunits. It interacts with the Smad transcription factors that are activated by TGF-β and prevents the heteromerization of Smad3 and Smad4. This results in inhibition of transcriptional activity of Smads and inhibition of TGF-β-induced differentiation of myofibroblasts. In sensory neurons, RGS3 mediates the termination of G protein signaling by calcium influx through voltage-gated channels. The role of RGS3 in collaboration with Ephrin-B is important in the maintenance of the neural progenitor cells.

Sequence

Synthetic peptide located within the following region: KDNLQSVTRGCFDLAQKRIFGLMEKDSYPRFLRSDLYLDLINQKKMSPPL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Douglas M Yau et al.
Molecular pharmacology, 73(5), 1356-1361 (2008-02-22)
Regulator of G protein signaling (RGS) proteins are united into a family by the presence of the homologous RGS domain that binds the alpha subunits of heterotrimeric G proteins and accelerates their GTPase activity. A member of this family, RGS3
Patrizia Tosetti et al.
Proceedings of the National Academy of Sciences of the United States of America, 100(12), 7337-7342 (2003-05-29)
G proteins modulate synaptic transmission. Regulators of G protein signaling (RGS) proteins accelerate the intrinsic GTPase activity of Galpha subunits, and thus terminate G protein activation. Whether RGS proteins themselves are under cellular control is not well defined, particularly in
Runxiang Qiu et al.
Stem cells (Dayton, Ohio), 28(9), 1602-1610 (2010-07-16)
Ephrin-B plays an important role in neural progenitor cells to regulate self-renewal and differentiation. Cellular and embryological evidence suggest this function of ephrin-B is mediated through a PDZ-dependent reverse signaling mechanism. Here, we have genetically investigated the function of PDZ-RGS3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service