Skip to Content
Merck
All Photos(2)

Key Documents

A9431

Sigma-Aldrich

Ancymidol

suitable for plant cell culture, BioReagent

Synonym(s):

reducymol, thritone, α-Cyclopropyl-α-(4-methoxyphenyl)-5-pyrimidinemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16N2O2
CAS Number:
Molecular Weight:
256.30
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Quality Level

form

powder

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(O)(C2CC2)c3cncnc3

InChI

1S/C15H16N2O2/c1-19-14-6-4-12(5-7-14)15(18,11-2-3-11)13-8-16-10-17-9-13/h4-11,18H,2-3H2,1H3

InChI key

HUTDUHSNJYTCAR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ancymidol, a-cyclopropyl-a-(4-methoxyphenyl)-5-pyrimidinemethanol (EL-53 1), a plant growth retardant is a synthetic pyrimidine analogue. It is also a weak fungitoxic.

Application

Ancymidol has been used as the cytochrome P450 inhibitor to study its effects on Cyanidioschyzon merolae strain.

Biochem/physiol Actions

Ancymidol is a pyrimidine-class plant cell endocrine disruptor plant growth regulator that interferes with gibberellin and cellulose biosynthesis.
Ancymidol acts as a monooxygenase inhibitor. It is capable of interrupting gibberellin biosynthesis and growth in plants.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Le Guen-Le Saos et al.
Annals of botany, 89(4), 419-425 (2002-07-05)
Bulbing was studied in shallot plants cultured in vitro. Bulbing occurred under a 16 h photoperiod with fluorescent + incandescent light and 30-50 g 1(-1) sucrose in the culture medium. Exogenous gibberellin (10 microM GA3) inhibited leaf and root growth
Hongyan Zheng et al.
Scientific reports, 6, 24778-24778 (2016-04-23)
The primary root plays essential roles in root development, nutrient absorption, and root architectural establishment. Primary root growth is generally suppressed by phosphate (P) deficiency in A. thaliana; however, the underlying molecular mechanisms are largely elusive to date. We found
Luisa M Trindade et al.
Gene, 303, 77-87 (2003-02-01)
In the search for time- and tissue-specific promoters an RNA fingerprinting technique called cDNA-AFLP was used. A transcript derived fragment (TDF511) was isolated which showed high similarity to alcohol dehydrogenases. The gene corresponding to this TDF, named Stgan, is likely
Marina Varbanova et al.
The Plant cell, 19(1), 32-45 (2007-01-16)
Arabidopsis thaliana GAMT1 and GAMT2 encode enzymes that catalyze formation of the methyl esters of gibberellins (GAs). Ectopic expression of GAMT1 or GAMT2 in Arabidopsis, tobacco (Nicotiana tabacum), and petunia (Petunia hybrida) resulted in plants with GA deficiency and typical
A E Desjardins et al.
Applied and environmental microbiology, 53(8), 1860-1865 (1987-08-01)
Ancymidol, a plant growth regulator, inhibited biosynthesis of diacetoxyscirpenol by Gibberella pulicaris (Fusarium sambucinum) in a defined liquid medium. Ancymidol also inhibited biosynthesis of T-2 toxin by a wild-type strain of Fusarium sporotrichioides and biosynthesis of diacetoxyscirpenol, deacetylated calonectrin, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service