Skip to Content
Merck
All Photos(1)

Key Documents

32523

Supelco

Flurprimidol

PESTANAL®, analytical standard

Synonym(s):

α-(1-Methylethyl)-α-[4-(trifluoromethoxy)phenyl]-5-pyrimidinemethanol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$239.00

$239.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$239.00

About This Item

Empirical Formula (Hill Notation):
C15H15F3N2O2
CAS Number:
Molecular Weight:
312.29
Beilstein:
892930
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$239.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)C(O)(c1ccc(OC(F)(F)F)cc1)c2cncnc2

InChI

1S/C15H15F3N2O2/c1-10(2)14(21,12-7-19-9-20-8-12)11-3-5-13(6-4-11)22-15(16,17)18/h3-10,21H,1-2H3

InChI key

VEVZCONIUDBCDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Flurprimidol is a plant growth retardant that finds application as a granular formulation.[1]

Application

Flurprimidol may be used as an analytical reference standard for the determination of the analyte in:
  • Vegetables and fruit samples by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction, followed by liquid chromatography (LC) coupled to full scan high resolution (Orbitrap) mass spectrometry (full scan HRMS).[1]
  • Tea samples by headspace solid-phase microextraction (HS-SPME) combined with gas chromatography–tandem mass spectrometry (GC–MS/MS).[2]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of a headspace solid-phase microextraction-based methodology followed by gas chromatography-tandem mass spectrometry for pesticide multiresidue determination in teas.
Li J, et al.
Chromatographia, 81(5), 809-821 (2018)
Timed release of flurprimidol from a granular formulation in mulches and sand.
Grey TL, et al.
Hortscience: a Publication of the American Society For Horticultural Science Hortscience, 44(2), 512-515 (2009)
Qualitative aspects and validation of a screening method for pesticides in vegetables and fruits based on liquid chromatography coupled to full scan high resolution (Orbitrap) mass spectrometry.
Mol HGJ, et al.
Analytical and Bioanalytical Chemistry, 403(10), 2891-2908 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service