Skip to Content
Merck
All Photos(1)

Documents

A6218

Sigma-Aldrich

Anti-ATM antibody, Mouse monoclonal

clone SYML6A10, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-ATM antibody produced in mouse, Anti-Ataxia-Telangiectasia Mutated

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SYML6A10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~350 kDa

species reactivity

human

technique(s)

immunocytochemistry: suitable
microarray: suitable
western blot: 1-2 μg/mL using total cell extract from a G-361 (human melanoma) cell line

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATM(472)

Looking for similar products? Visit Product Comparison Guide

General description

Ataxia-Telangiectasia Mutated (ATM) is a member of the phosphatidylinositol 3-kinase (PI-3) family of kinases. It is activated in response to DNA damage such as double stranded breaks.
Mouse Monoclonal Anti-ATM antibody binds to human ATM.

Immunogen

synthetic peptide corresponding to the C-terminal region of human ATM with N-terminally added cysteine, conjugated to KLH.

Application

Mouse Monoclonal Anti-ATM antibody can be used for western blot assays at 1-2μg/ml using G-361 cell extracts. The antibody can also be used for immunocytochemistry and microarray applications.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Yang et al.
Carcinogenesis, 24(10), 1571-1580 (2003-08-16)
Exposure to genotoxic agents is a major cause of human cancer, and cellular responses to genotoxic stress are important defense mechanisms. These responses are very complex, involving many cellular factors that form an extensive signal transduction network. This network includes
Karen S Yee et al.
Cancer research, 72(9), 2206-2217 (2012-03-06)
RASSF1A (Ras association domain containing family 1A), a tumor suppressor gene that is frequently inactivated in human cancers, is phosphorylated by ataxia telangiectasia mutated (ATM) on Ser131 upon DNA damage, leading to activation of a p73-dependent apoptotic response. A single-nucleotide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service