Skip to Content
Merck
All Photos(1)

Key Documents

A5883

Sigma-Aldrich

8-Azaadenine

≥99%

Synonym(s):

6-Amino-8-azapurine, 8-Aza-6-aminopurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N6
CAS Number:
Molecular Weight:
136.11
Beilstein:
141551
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥99%

form

powder

mp

>300 °C (lit.)

solubility

1 M NaOH: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

Nc1ncnc2nn[nH]c12

InChI

1S/C4H4N6/c5-3-2-4(7-1-6-3)9-10-8-2/h1H,(H3,5,6,7,8,9,10)

InChI key

HRYKDUPGBWLLHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Base pairs of azaadenine with thymine (and azaguanine with cytosine) are found by ab initio calculation to be up to 7 kcal/mol more stable than natural AT (and GC) base pairs.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Biagi et al.
Farmaco (Societa chimica italiana : 1989), 50(1), 13-19 (1995-01-01)
Comparison of the affinity towards adenosine receptors of 2-phenyl-8-azaadenosines, bearing a lipophilic substituent on N(6), with the corresponding 2-phenyl-8-azaadenines was carried out. The compounds have good A1 affinity and high A1 selectivity. The obtained Ki(rib)/Ki(benz) ratios for A1 receptors, which
D J Chen et al.
Environmental mutagenesis, 4(4), 457-467 (1982-01-01)
A chinese hamster ovary (CHO) cell line heterozygous at the adenine phosphoribosyl transferase (APRT) locus was used for selection of induced mutants resistant to 8-azaadenine (8AA), 6-thioguanine (6TG), ouabain (OUA), emetine (EMT) and diphtheria toxin (DIP). The expression times necessary
M I Nieto et al.
Nucleosides & nucleotides, 17(7), 1255-1266 (1998-08-26)
Six new carbocyclic nucleosides were prepared by mounting a purine (compounds 5-7), 8-azapurine (compounds 9 and 10) or pyrimidine (compound 13) base on the amino group of (1R,cis)-3-(aminomethyl)-1,2,2-trimethylcyclopentylmethanol (2). The antiviral activity of compounds 5-7, 10 and 13, and their
A E Simon et al.
Molecular and cellular biology, 3(10), 1703-1710 (1983-10-01)
A two-step model to explain the high frequency of mutation at the diploid adenine phosphoribosyltransferase (aprt) locus in CHO cells has been proposed previously (Simon et al., Mol. Cell. Biol. 2:1126-1133, 1982). This model indicates that two distinct classes of
Giuliana Biagi et al.
Farmaco (Societa chimica italiana : 1989), 57(3), 221-233 (2002-05-07)
erythro-2-Phenyl-9-(2-hydroxy-3-nonyl)adenine and its 8-aza analog were prepared and showed a very high inhibitory activity towards adenosine deaminase (ADA), with Ki 0.55 and 1.67 nM, respectively, and high affinity for A1 adenosine receptors, with Ki 28 and 2.8 nM, respectively. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service