Skip to Content
Merck
All Photos(1)

Key Documents

A4233

Sigma-Aldrich

Ara-G hydrate

≥98% (HPLC), solid

Synonym(s):

9-β-D-Arabinofuranosyl guanine hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$184.00
25 MG
$699.00

$184.00


Estimated to ship onMay 20, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$184.00
25 MG
$699.00

About This Item

Empirical Formula (Hill Notation):
C10H13N5O5 · xH2O
CAS Number:
Molecular Weight:
283.24 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$184.00


Estimated to ship onMay 20, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: >10 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

NC1=Nc2c(ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O)C(=O)N1

InChI

1S/C10H13N5O5/c11-10-13-7-4(8(19)14-10)12-2-15(7)9-6(18)5(17)3(1-16)20-9/h2-3,5-6,9,16-18H,1H2,(H3,11,13,14,19)/t3-,5-,6+,9-/m1/s1

InChI key

NYHBQMYGNKIUIF-FJFJXFQQSA-N

Related Categories

Application

Ara-G is converted by cellular kinases to the active 5′-triphosphate, Ara-GTP. This active form of Ara-G induces apoptosis and inhibits DNA synthesis. Ara-G is also an antineoplastic and antimetabolite.

Biochem/physiol Actions

Ara-G is an inducer of apoptosis; inhibitor of DNA synthesis; antineoplastic; and antimetabolite.
Ara-G is an inducer of apoptosis; inhibitor of DNA synthesis; antineoplastic; and antimetabolite. Ara-G is converted by cellular kinases to the active 5′-triphosphate, Ara-GTP. Incorporation of Ara-GTP into DNA leads to inhibition of DNA synthesis and apoptosis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Yamauchi et al.
Oncology reports, 23(2), 499-504 (2010-01-01)
The 9-beta-D-arabinofuranosylguanine (ara-G), an active compound of nelarabine, demonstrates potent cytotoxicity specifically on T-cell malignancies. In cells, ara-G is phosphorylated to ara-G triphosphate (ara-GTP), which is subsequently incorporated into DNA, thereby inhibiting DNA synthesis. Because ara-GTP is crucial to ara-G's
M Bjerke et al.
Nucleosides, nucleotides & nucleic acids, 27(6), 746-749 (2008-07-05)
Incubation of cells with thymidine (dThd) is known to cause dNTP pool imbalance as well as deletions and depletion of the mtDNA. In order to gain further understanding in the events involved in dThd toxicity over time, H9 cells were
A Fyrberg et al.
Biochemical and biophysical research communications, 427(3), 456-460 (2012-08-23)
Our previous data from a human leukemic cell line made resistant to the nucleoside analog (NA) 9-β-D-arabinofuranosylguanine (AraG) revealed a massive upregulation of fetal hemoglobin (HbF) genes and the ABCB1 gene coding for the multidrug resistance P-glycoprotein (P-gp). The expression
Anna Fyrberg et al.
Cancer chemotherapy and pharmacology, 68(3), 583-591 (2010-11-27)
To characterize resistance mechanisms to the nucleoside analog 9-β-D-arabinofuranosylguanine (AraG) in the T-cell acute lymphoblastic leukemia cell line MOLT-4 and its AraG-resistant variant. A gene expression microarray analysis was performed, as well as gene expression and enzyme activity measurements of
Sheryl A Flanagan et al.
Biochemical pharmacology, 66(5), 733-737 (2003-09-02)
We studied acceptance of various deoxyguanosine analogues by the unique guanosine preferring nucleoside transport system exhibited by NB4 cells, csg. Indirect assessment of acceptance using transport inhibition assays revealed that both 1-beta-D-arabinofuranosylguanine (ara-G) and 4'-thio-beta-D-xylofuranosylguanine (thio-xyl-G) compete with guanosine for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service