Skip to Content
Merck
All Photos(1)

Key Documents

A3111

Sigma-Aldrich

A1120

≥98% (HPLC), powder

Synonym(s):

2-(4-(2-(trifluoromethyl)phenyl)piperidine-1-carboxamido)benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$128.00
25 MG
$600.00

$128.00


Estimated to ship onApril 11, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$128.00
25 MG
$600.00

About This Item

Empirical Formula (Hill Notation):
C20H19F3N2O3
CAS Number:
Molecular Weight:
392.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$128.00


Estimated to ship onApril 11, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

off-white

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

OC(=O)c1ccccc1NC(=O)N2CCC(CC2)c3ccccc3C(F)(F)F

InChI

1S/C20H19F3N2O3/c21-20(22,23)16-7-3-1-5-14(16)13-9-11-25(12-10-13)19(28)24-17-8-4-2-6-15(17)18(26)27/h1-8,13H,9-12H2,(H,24,28)(H,26,27)

InChI key

MEAQCLPMSVEOQF-UHFFFAOYSA-N

Biochem/physiol Actions

A1120 is a selective non-retinoid ligand for retinol-binding protein 4 (RBP4).
A1120 is a selective non-retinoid ligand for retinol-binding protein 4 (RBP4). RBP4 transports retinol from the liver to extrahepatic tissues and RBP4 lowering is reported to improve insulin sensitivity in mice. A1120 is a high affinity non-retinoid ligand for RBP4 which disrupts the interaction between RBP4 and its binding partner transthyretin (TTR). It binds to the same site as retinol and induces changes in the orientation (compared to the retinol bound form) of loops at the RBP4-TTR interaction interface.A1120 lowers RBP4 and retinol levels in a dose-dependent manner, to a similar extent as seen with fenretinide. However, unlike fenretinide (Sigma# H7779), A1120 does not have beneficial effects on insulin resistance.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebecca Ward et al.
The Journal of biological chemistry, 295(19), 6482-6497 (2020-04-03)
Cone photoreceptors in the retina enable vision over a wide range of light intensities. However, the processes enabling cone vision in bright light (i.e. photopic vision) are not adequately understood. Chromophore regeneration of cone photopigments may require the retinal pigment
Astrid M Bedoya et al.
Gynecologic oncology, 135(1), 74-80 (2014-08-02)
Cervical cancer is characterized by an immunosuppressive microenvironment and a Th2-type cytokine profile. Expression of arginase (ASE), the enzyme that converts L-arginine into L-ornithine and urea, is stimulated by Th2-type cytokines. To assess the association of ASE activity and L-Arg
Jaume Amengual et al.
Human molecular genetics, 23(20), 5402-5417 (2014-05-24)
Vitamin A must be adequately distributed within the body to maintain the functions of retinoids in the periphery and chromophore production in the eyes. Blood transport of the lipophilic vitamin is mediated by the retinol-binding protein, RBP4. Biochemical evidence suggests
Esteban A Lopera et al.
Infection, genetics and evolution : journal of molecular epidemiology and evolutionary genetics in infectious diseases, 28, 339-348 (2014-12-03)
European (E) variants of HPV 16 are evenly distributed among world regions, meanwhile Non-European variants such as European-Asian (EAs), Asian American (AA) and African (Af) are mostly confined to Eastern Asia, The Americas and African regions respectively. Several studies have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service