SIAL0167
Sigma® cell culture dish
diam. × H 100 mm × 20 mm, tissue-culture treated, sterile, pack of 25 × 20 ea
All Photos(1)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.22
Recommended Products
sterility
sterile
packaging
pack of 25 × 20 ea
diam. × H
100 mm × 20 mm
surface area
55 cm2
binding type
Tissue Culture (TC)-treated surface
General description
- Manufactured from optically-clear virgin polystyrene
- Treated for optimal cell attachment
- Sterilized by gamma radiation and certified nonpyrogenic
- Have stacking beads to aid in handling
- Lid supplied with vents to provide consistent gas exchange
Legal Information
Sigma is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Already Own This Product?
Customers Also Viewed
Protocols
Universal Transfection Reagent enables efficient nucleic acid delivery into various cells, compatible with different cell culture conditions.