Key Documents
SIAL0166
Sigma-Aldrich
Sigma® cell culture dish
diam. × H 60 mm × 15 mm, tissue-culture treated, sterile, pack of 25 × 20 ea
About This Item
UNSPSC Code:
41121812
NACRES:
NB.22
sterility
sterile
packaging
pack of 25 × 20 ea
diam. × H
60 mm × 15 mm
surface area
21 cm2
binding type
Tissue Culture (TC)-treated surface
General description
  • Manufactured from optically-clear virgin polystyrene
  • Treated for optimal cell attachment
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Lid supplied with vents to provide consistent gas exchange
Legal Information
Sigma is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
Store the cryovials in a liquid nitrogen storage tank immediately upon arrival.
Technical information for working with bovine aortic endothelial cells including thawing, subculturing and cryopreservation.
Culturing HCFs from human heart ventricles involves critical steps for studying cardiac-related conditions.
Technical information for working with human aortic endothelial cells including thawing, subculturing and cryopreservation. Be sure to wear face protection mask and gloves when retrieving cryovials from the liquid nitrogen storage tank. The dramatic temperature change from the tank to the room could cause any trapped liquid nitrogen in the cryovials to burst and cause injury.
See All