Skip to Content
Merck
All Photos(1)

Key Documents

96063

Sigma-Aldrich

Coumestrol dimethyl ether

BioReagent, suitable for fluorescence, ≥95% (HPLC)

Synonym(s):

7,12-Dimethoxy-coumestan

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$349.00

$349.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$349.00

About This Item

Empirical Formula (Hill Notation):
C17H12O5
CAS Number:
Molecular Weight:
296.27
Beilstein:
286103
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

$349.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

product line

BioReagent

Assay

≥95% (HPLC)

form

powder

solubility

DMSO: soluble

fluorescence

λex 325 nm; λem 421 nm in 0.1 M Tris pH 8.0
λex 377 nm; λem 437 nm in 0.1 M Tris pH 8.0 (reaction product, free coumetrol)

suitability

suitable for fluorescence

SMILES string

COc1ccc-2c(OC(=O)c3c-2oc4cc(OC)ccc34)c1

InChI

1S/C17H12O5/c1-19-9-3-5-11-13(7-9)21-16-12-6-4-10(20-2)8-14(12)22-17(18)15(11)16/h3-8H,1-2H3

InChI key

PXHLPCBBXPHBHP-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service